Potassium chloride 99
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Potassiumchloride
A11662.0B A11662.0G A11662.0I
A11662.0BEA 40.1 EUR
A11662.0B A11662.0G A11662.0I
Potassium chloride 99
Potassium chloride
Purity: 99% CAS No.: 7447-40-7 Molecular Formula: KCl Formula Weight: 74.55 Refractive Index: 1.490 Odour: Odourless Boiling Point: 1500° subl. Merck Reference: 14,7621 MDL No.: MFCD00011360
Formule: KCl
MW: 74.55 g/mol
Kookpunt: 1500 °C (sublimatie)
Smeltpunt: 773
Dichtheid: 1.984
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011360
CAS nummer: 7447-40-7
EINECS: 231-211-8
Merck Index: 14,7621