Lithiumchloride, anhydrous 99%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
10515.30 10515.A1
10515.30EA 92.5 EUR
10515.30 10515.A1
Lithiumchloride, anhydrous 99%
Lithium chloride
Purity: 99% CAS No.: 7447-41-8 Molecular Formula: ClLi Formula Weight: 42.39 Physical Form: -20 Mesh Density: 2.068 Appearance: White Odour: Odourless Melting Point: 605° Boiling Point: 1325-1360° Merck Reference: 14,5528 MDL No.: MFCD00011078
Formule: LiCl
MW: 42.39 g/mol
Kookpunt: 1325…1360 °C
Smeltpunt: 605 °C
Dichtheid: 2.068
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011078
CAS nummer: 7447-41-8
EINECS: 231-212-3
Merck Index: 14,5528