Natriumchloride, Optical Grade Optical Grade
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
|
Synoniemen:
Saline, NaCl, Sea salt, Halite
11019.18
11019.30
11019.18EA
129
EUR
11019.18
11019.30
Natriumchloride, Optical Grade Optical Grade
Natriumchloride
CAS No.: 7647-14-5
Molecular Formula: NaCl
Formula Weight: 58.44
Physical Form: Broken Crystals
Density: 2.165
Refractive Index: 1.5442
Sensitivity: Hygroscopic
Appearance: White
Odour: Odourless
pH: 6-9
Melting Point: 801°
Boiling Point: 1413°
Merck Reference: 14,8599
MDL No.: MFCD00003477
Formule:
NaCl MW: 58.44 g/mol Kookpunt: 1413 °C Smeltpunt: 801 Dichtheid: 2.165 Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00003477 CAS nummer: 7647-14-5 EINECS: 231-598-3 Merck Index: 14,8599 |