Natriumchloride 99+%, crystalline powder PDV
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Saline, NaCl, Sea salt, Halite
A12313.0B A12313.0G A12313.0I
A12313.0BEA 24 EUR
A12313.0B A12313.0G A12313.0I
Natriumchloride 99+%, crystalline powder PDV
Natriumchloride
Purity: 99+% CAS No.: 7647-14-5 Molecular Formula: NaCl Formula Weight: 58.44 Refractive Index: 1.5442 Odour: Odourless pH: 6-9 Melting Point: 801° Boiling Point: 1413° Merck Reference: 14,8599 MDL No.: MFCD00003477
Formule: NaCl
MW: 58.44 g/mol
Kookpunt: 1413 °C
Smeltpunt: 801
Dichtheid: 2.165
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003477
CAS nummer: 7647-14-5
EINECS: 231-598-3
Merck Index: 14,8599