Overhead stirrer tools, impellers, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
C389-28 C389-32 C389-66 C389-20 C389-18 C389-62 C389-36 C389-22
BOHLC389-28EA 437 EUR
BOHLC389-28 BOHLC389-32 BOHLC389-66 BOHLC389-20 BOHLC389-18 BOHLC389-62 BOHLC389-36 BOHLC389-22
Overhead stirrer tools, impellers, BOLA
Roerders Roergereedschap
Stirrer shafts made of PTFE-jacketed stainless steel and the impeller completely made of PTFE with three blades bent backwards.
  • Suitable for stirring in vessels with round bottom
  • Resistant to chemicals
  • Temperature resistance from –200 to +250 °C