Overhead stirrer tools, radial stirrers, six paddles, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
C598-12 C598-26 C598-32 C598-22 C598-16 C598-46 C598-36 C598-42 C598-56 C598-18
BOHLC598-12EA 215 EUR
BOHLC598-12 BOHLC598-26 BOHLC598-32 BOHLC598-22 BOHLC598-16 BOHLC598-46 BOHLC598-36 BOHLC598-42 BOHLC598-56 BOHLC598-18
Overhead stirrer tools, radial stirrers, six paddles, BOLA
Roerders Roergereedschap
Stirrer shafts made of PTFE-jacketed stainless steel and the impeller completely made of PTFE with six radial paddles.
  • Resistant to chemicals
  • Temperature resistance from –200 to +250 °C
Axial suction of mixture, strong radial flow. Ideal for aerating liquids.