Bottles, wide neck, with screw cap, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A100-01 A100-08 A100-04 A100-07 A100-06 A100-03 A100-10 A100-09 A100-05
BOHLA100-01EA 19.6 EUR
BOHLA100-01 BOHLA100-08 BOHLA100-04 BOHLA100-07 BOHLA100-06 BOHLA100-03 BOHLA100-10 BOHLA100-09 BOHLA100-05
Bottles, wide neck, with screw cap, BOLA
Flessen Kunststof flessen
PTFE, white.
  • Temperature resistance: from −200 to +250 °C
  • Universal chemical resistance
  • Pressure: no pressure
Thick-walled, smooth interior surface, with screw cap.