Bottles, wide neck, PFA, conical neck, with screw cap, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A103-09 A103-12 A103-15 A103-03 A103-06
BOHLA103-09EA 235 EUR
BOHLA103-09 BOHLA103-12 BOHLA103-15 BOHLA103-03 BOHLA103-06
Bottles, wide neck, PFA, conical neck, with screw cap, BOLA
Flessen Kunststof flessen
PFA, translucent.
  • Temperature resistance: from −200 to +250 °C
  • Universal chemical resistance
  • Pressure: no pressure
  • Non porous
Conical neck with screw cap.