Magnetic stirrer heads, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
C512-08 C502-08
BOHLC512-08EA 1450 EUR
BOHLC512-08 BOHLC502-08
Magnetic stirrer heads, BOLA
Roerkoppen
Magnetic stirrer heads consist of perfect combination of drive shaft with ball bearings, rotor and lower bearing made of PTFE and a conductor made of borosilicate glass.
  • No grease required
  • Safe to run dry
  • Long durability and compact size
  • No leakage or memory effects due to non-porous, welded rotor
  • Temperature range: −100 to 240 °C
  • Chemical resistance
The rotor of this unit holds the stirrer shaft by means of three stud screws which are fixed in the counterbores of the stirrer shaft. The 6 mm square can be fixed into the stirrer coupling or into the agitator.
The outer Ø of space saving drive shaft is measured as 28 mm and 38 mm for NS 19/26 and NS 29/32 or NS 45/40 respectively.