Jars, with screw cap, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A130-05 A130-07 A130-01 A130-12 A130-11 A130-09 A130-06
BOHLA130-05EA 161 EUR
BOHLA130-05 BOHLA130-07 BOHLA130-01 BOHLA130-12 BOHLA130-11 BOHLA130-09 BOHLA130-06
Jars, with screw cap, BOLA
Potjes Potjes voor algemeen gebruik
Made of PFA, transparent with screw-cap.
  • Sturdy design
  • Non-porous
  • Good chemical resistance
  • Temperature resistance from −200 to +250 °C