Jars, with screw cap and tubing connections, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A131-12 A131-14
BOHLA131-12EA 389 EUR
BOHLA131-12 BOHLA131-14
Jars, with screw cap and tubing connections, BOLA
Potjes Potjes voor algemeen gebruik
Jars, made of PFA, transparent with screw-cap.
  • Sturdy design
  • Non-porous
  • Good chemical resistance
  • Temperature resistance from −200 to +250 °C
  • FDA conform
Tubings with an external diameter of max. 6,35 mm can be inserted through the screw caps.