GL-screw-in tube fittings
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
D516-50 D516-44 D517-32 D517-14 D517-38 D516-32 D517-26 D517-08 D516-38 D516-26 D516-20 D516-14 D516-08 D517-50 D517-20 D517-74
BOHLD516-50EA 81.7 EUR
BOHLD516-50 BOHLD516-44 BOHLD517-32 BOHLD517-14 BOHLD517-38 BOHLD516-32 BOHLD517-26 BOHLD517-08 BOHLD516-38 BOHLD516-26 BOHLD516-20 BOHLD516-14 BOHLD516-08 BOHLD517-50 BOHLD517-20 BOHLD517-74
GL-screw-in tube fittings
Fittingen en connectoren
Straight tube fitting, made of PTFE, with one screw-in thread (either NPT or G thread).
  • Universal chemical resistance
  • Temperature resistance from -200 up to +250 °C
  • Pressure: 10 bar
  • Suitable for vacuum