Virginal rods, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
S1800-06 S1800-12 S1800-25 S1800-30 S1800-40 S1800-20 S1800-16 S1800-15 S1800-10 S1800-08
BOHLS1800-06EA 38.1 EUR
BOHLS1800-06 BOHLS1800-12 BOHLS1800-25 BOHLS1800-30 BOHLS1800-40 BOHLS1800-20 BOHLS1800-16 BOHLS1800-15 BOHLS1800-10 BOHLS1800-08
Virginal rods, BOLA
Statiefstaven
Virginal rods made of white PTFE.
  • Diameter and length are nominal dimensions and can contain a machining allowance
  • Temperature resistance from −200 to +250 °C
  • Universal chemical resistance
  • FDA conform