Sampling bags for the storage or gases and liquids, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A223-62 A223-74 A223-70 A223-58
BOHLA223-62EA 63.4 EUR
BOHLA223-62 BOHLA223-74 BOHLA223-70 BOHLA223-58
Sampling bags for the storage or gases and liquids, BOLA
Zakken Monsterzakken
These PVF inert sampling bags are transparent, non-porous and are without plasticisers that are suitable for the storage of gas and liquids.
  • With good chemical resistance
  • Temperature resistance from -200 to +250 °C
Leveringsinformatie: Supplied with two exchangeable cylindrical ports made of PTFE.