Connectors for sampling bags, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A226-12 A226-06
BOHLA226-12EA 35.6 EUR
BOHLA226-12 BOHLA226-06
Connectors for sampling bags, BOLA
Fittingen en connectoren
Made of PTFE, white
  • Temperature resistance: from −200 to +250°C
  • Universal chemical resistance:
Connectors to be screwed in the ports of sampling bags with bore Ø 3 mm and outer Ø 6 mm.
Certificaten: FDA conform.