Dropping funnels, conical, FEP, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
B285-03 B285-02 B285-01
BOHLB285-03EA 1160 EUR
BOHLB285-03 BOHLB285-02 BOHLB285-01
Dropping funnels, conical, FEP, BOLA
Trechters Druppeltrechters
Dropping funnel with fine adjustment stopcock having cone size 29 made of PTFE.
  • Temperature resistance from 0 to +110 °C
  • Universal chemical resistance
  • Transparent
With integrated special nut for easy locking and unlocking of the ground joint.
Certificaten: FDA conform.