Culture bottles, for magnetic stirring, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
C420-03 C420-05
BOHLC420-03EA 255 EUR
BOHLC420-03 BOHLC420-05
Culture bottles, for magnetic stirring, BOLA
Kolven Cultuurflessen
Culture bottles, made of borosilicate glass, suitable for both low and high speed (maximum 1000 rpm) stirrers for gentle mixing of cell cultures.
  • Universal chemical resistance
  • Screw caps for sidearms made of PPS
  • Screw cover for center neck made of PP with glass fibre
Stirrer made of PTFE and stainless steel is continuously adjustable in height from the outside. Stirring unit is driven by a common magnetic stirrer and magnetism causes rotation. Sidearms can be connected to tubing, probes or sensors.