Overhead stirrer tools, paddle stirrers, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
C379-02 C379-08 C379-10 C379-18 C379-12 C379-06 C379-04
BOHLC379-02EA 288 EUR
BOHLC379-02 BOHLC379-08 BOHLC379-10 BOHLC379-18 BOHLC379-12 BOHLC379-06 BOHLC379-04
Overhead stirrer tools, paddle stirrers, BOLA
Roerders Roergereedschap
Stirrer shafts made of PTFE-jacketed stainless steel and the impeller completely made of PTFE with two 45° angled blades.
  • Creates axial flow
  • Resistant to chemicals
  • Temperature resistance from –200 to +250 °C