Powder pillows, Chloride 2 indicator
Leverancier: Hach
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
104399 105766
HACH104399EA 57.8 EUR
HACH104399 HACH105766
Powder pillows, Chloride 2 indicator
Milieutestkits
Powder reagent in sealed package used as indicator for the argentometric titration of chloride.
Opgelet: Note: Product contains potsassium chromate. Please read MSDS prior to usage.