Kaliumfluoride, anhydrous 99%, extra pure
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
201350010 201350250 201352500 201355000 201350025
ACRO201350010EA 71.9 EUR
ACRO201350010 ACRO201350250 ACRO201352500 ACRO201355000 ACRO201350025
Kaliumfluoride, anhydrous 99%, extra pure
Kaliumfluoride
Formule: KF
Kookpunt: 1505 °C (1013 hPa)
Smeltpunt: 860 °C
Dichtheid: 2,48 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011398
CAS nummer: 7789-23-3
EINECS: 232-151-5
UN: 1812
ADR: 6.1,III
Merck Index: 13,07714
Order Now
Specification Test Results
Appearance White Crystalline powder or crystals and/or chunks
Titration ≥98.5 %
Chloride (Cl) ≤500 ppm
Sulfate (SO4) ≤1000 ppm
Iron (Fe) ≤50 ppm