Lithium fluoride, ultra dry ≥99.99% (metals basis)
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Ampuled under argon
14463.09
14463.18
14463.09EA
212
EUR
14463.09
14463.18
Lithium fluoride, ultra dry ≥99.99% (metals basis)
Lithium fluoride
Purity: 99.99%
CAS No.: 7789-24-4
Molecular Formula: FLi
Formula Weight: 25.94
Physical Form: Powder, Ampouled under argon
Density: 2.635
Refractive Index: 1.3915
Sensitivity: Hygroscopic
Appearance: White
Odour: Odourless
Melting Point: 845°
Boiling Point: 1680°
Merck Reference: 14,5531
UN No.: 3288
MDL No.: MFCD00011090
Formule:
FLi Kookpunt: 1680 °C Smeltpunt: 845 °C Dichtheid: 2,635 Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00011090 CAS nummer: 7789-24-4 EINECS: 232-152-0 UN: 3288 ADR: 6.1,III Merck Index: 14,5531 |