Potassium iodide 99.995% (trace metals basis), extra pure
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
193790500 193790100 193792500
ACRO193790500EA 239 EUR
ACRO193790500 ACRO193790100 ACRO193792500
Potassium iodide 99.995% (trace metals basis), extra pure
Potassium iodide
Formule: Kl
Kookpunt: 1330 °C (760 mmHg)
Smeltpunt: 680 °C
Dichtheid: 3,13 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011405
CAS nummer: 7681-11-0
EINECS: 231-659-4
Merck Index: 13,07727
Order Now
Specification Test Results
Appearance White Powder or crystals
Total trace metal impurities ≤0.005 %