Potassium iodide ≥99.9% (metals basis)
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
Dipotassium diiodide, Kali iodide
42857.14
42857.22
42857.36
42857.14EA
48.8
EUR
42857.14
42857.22
42857.36
Potassium iodide ≥99.9% (metals basis)
Potassium iodide
Purity: 99.9%
CAS No.: 7681-11-0
Molecular Formula: KI
Formula Weight: 166.01
Physical Form: Powder
Density: 3.12
Refractive Index: 1.677
Sensitivity: Hygroscopic
Odour: Odourless
Melting Point: 681°
Boiling Point: 1330°
Merck Reference: 14,7643
MDL No.: MFCD00011405
Formule:
Kl Kookpunt: 1330 °C Smeltpunt: 681 °C Dichtheid: 3,12 Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00011405 CAS nummer: 7681-11-0 EINECS: 213-659-4 Merck Index: 14,7643 |