Lithium iodide, ultra dry ≥99.999% (metals basis)
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Ampuled under argon
13600.06
13600.14
13600.22
13600.06EA
142
EUR
13600.06
13600.14
13600.22
Lithium iodide, ultra dry ≥99.999% (metals basis)
Lithium iodide
Purity: 99.999%
CAS No.: 10377-51-2
Molecular Formula: LiI
Formula Weight: 133.84
Physical Form: -10 Mesh Beads, Ampouled under argon
Density: 4.076
Refractive Index: 1.955
Sensitivity: Light Sensitive and Hygroscopic
Appearance: White to off-white
Odour: Odourless
Melting Point: 449°
Boiling Point: 1170-1190°
Merck Reference: 14,5535
MDL No.: MFCD00011092
Formule:
ILi Kookpunt: 1170…1190 °C Smeltpunt: 449 °C Dichtheid: 4,076 Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00011092 CAS nummer: 10377-51-2 EINECS: 233-822-5 Merck Index: 14,5535 |