Lithium iodide, watervrij 98+%
Leverancier: Thermo Fisher ScientificTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Ampuled under argon
Purity: 98+%
CAS No.: 10377-51-2
Molecular Formula: ILi
Formula Weight: 133.84
Physical Form: Lumps/powder, Ampouled under argon
Density: 4.076
Refractive Index: 1.955
Sensitivity: Light Sensitive and Hygroscopic
Appearance: White to pale yellow
Odour: Odourless
Melting Point: 449°
Boiling Point: 1170-1190°
Solubility: Soluble in water, alcohol, acetone and methanol. Very soluble in NH4OH
Merck Reference: 14,5535
MDL No.: MFCD00011092
As absorbent in refrigeration process, as a catalyst, and in photography
Formule:
ILi Kookpunt: 1170…1190 °C Smeltpunt: 449 °C Dichtheid: 4,076 Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00011092 CAS nummer: 10377-51-2 EINECS: 233-822-5 Merck Index: 14,5535 |