Sodium iodide, anhydrous 99+%, pure
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Hydriodic acid sodium salt, Sodium monoiodide, Ioduril
203180010 203180050 203182500 203180025 203180100
ACRO203180010EA 275 EUR
ACRO203180010 ACRO203180050 ACRO203182500 ACRO203180025 ACRO203180100
Sodium iodide, anhydrous 99+%, pure
Sodium iodide
Formule: NaI
Kookpunt: 1300 °C (760 mmHg)
Smeltpunt: 661 °C
Dichtheid: 3,66 g/cm³
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003532
CAS nummer: 7681-82-5
EINECS: 231-679-3
UN: 3077
ADR: 9,III
Merck Index: 13,08705
Order Now
Specification Test Results
Appearance White Crystalline powder
Titration Argentometric ≥99.0 % (on dried substance)
Loss on drying ≤3 % (1 g, 105°C, 3 h)