Potassium iodate FCC, extra pure, SAFC®
Leverancier: MERCK PRODUCTION CHEMICALS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
1.05050.0500 1.05050.9029
1.05050.0500EA 531 EUR
1.05050.0500 1.05050.9029
Potassium iodate FCC, extra pure, SAFC®
Potassium iodate
Formule: KlO₃
MW: 214 g/mol
Smeltpunt: 560
Dichtheid: 3.98 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011406
CAS nummer: 7758-05-6
EINECS: 231-831-9
UN: 1479
ADR: 5.1 II
Merck Index: 13,07726