Kaliumpermanganaat ≥99%, Puriss. p.a. Reag. Ph. Eur., ACS analytical reagent, Fluka™
Leverancier: Honeywell Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Permanganic acid potassium salt
31404-100G 31404-1KG 31404-2.5KG 31404-500G 31404-25KG
31404-100GEA 37.9 EUR
31404-100G 31404-1KG 31404-2.5KG 31404-500G 31404-25KG
Kaliumpermanganaat ≥99%, Puriss. p.a. Reag. Ph. Eur., ACS analytical reagent, Fluka™
Kaliumpermanganaat
Formule: KMnO₄
MW: 158.03 g/mol
Smeltpunt: Dec.
Dichtheid: 2.700 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011364
CAS nummer: 7722-64-7
EINECS: 231-760-3
UN: 1490
ADR: 5.1 II
Merck Index: 12,07824
Order Now
Specification Test Results
Assay Min. 99.0 %
Water insoluble matter Max. 0.2 %
Loss on drying (105°C, 2 h) Max. 0.2 %
Chloride (Cl) Max. 200 ppm
Chloride, Chlorate (as Cl) Max. 0.005 %
Sulfate (SO₄) Max. 200 ppm
Appearance of the solution complying