Sodium molybdate(VI) Mo 46,2%, watervrij
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
12209.22
12209.36
12209.22EA
51.8
EUR
12209.22
12209.36
Sodium molybdate(VI) Mo 46,2%, watervrij
Sodium molybdate(VI)
CAS No.: 7631-95-0
Molecular Formula: MoNa2O4
Formula Weight: 205.92
Physical Form: Powder
Density: 3.72
Refractive Index: 1.714
Sensitivity: Hygroscopic
Appearance: White
Odour: Odourless
Melting Point: 687°
Merck Reference: 14,8645
MDL No.: MFCD00003486
Formule:
MoNa₂O₄ Smeltpunt: 687 °C Dichtheid: 3,72 Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00003486 CAS nummer: 7631-95-0 EINECS: 231-551-7 Merck Index: 14,8645 |