Cesium nitrate ≥99.8% (metals basis)
Leverancier: Thermo Fisher ScientificTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
Nitric acid cesium salt
Purity: 99.8%
CAS No.: 7789-18-6
Molecular Formula: CsNO3
Formula Weight: 194.91
Physical Form: Crystalline
Density: 3.68
Refractive Index: 1.55
Sensitivity: Hygroscopic
Appearance: White
Odour: Odourless
Melting Point: 414°
Boiling Point: dec.
Solubility: Soluble in 5 parts cold and 0.5 parts boiling water. Soluble in acetone. Slightly soluble in alcohol
Merck Reference: 14,2015
UN No.: 1451
MDL No.: MFCD00010963
Preparation of cesium compounds
Formule:
CsNO₃ MW: 194.91 g/mol Kookpunt: 737 Dec. Smeltpunt: 414 Dichtheid: 3.68 Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00010963 CAS nummer: 7789-18-6 EINECS: 232-146-8 UN: 1451 ADR: 5.1 III Merck Index: 14,2015 |