Cesium nitrate ≥99.99% (metals basis)
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
Nitric acid cesium salt
14440.06
14440.14
14440.22
14440.36
14440.06EA
35.3
EUR
14440.06
14440.14
14440.22
14440.36
Cesium nitrate ≥99.99% (metals basis)
Cesium nitrate
Purity: 99.99%
CAS No.: 7789-18-6
Molecular Formula: CsNO3
Formula Weight: 194.91
Physical Form: Crystalline
Density: 3.68
Refractive Index: 1.55
Sensitivity: Hygroscopic
Appearance: White
Odour: Odourless
Melting Point: 414°
Boiling Point: dec.
Merck Reference: 14,2015
UN No.: 1451
MDL No.: MFCD00010963
Formule:
CsNO₃ MW: 194.91 g/mol Kookpunt: 737 Dec. Smeltpunt: 414 Dichtheid: 3.68 Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00010963 CAS nummer: 7789-18-6 EINECS: 232-146-8 UN: 1451 ADR: 5.1 III Merck Index: 14,2015 |