Potassium nitrate 99,995 Metals basis, Puratronic®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Nitrate of Potash
10844.06 10844.18 10844.30
10844.06EA 52.6 EUR
10844.06 10844.18 10844.30
Potassium nitrate 99,995 Metals basis, Puratronic®
Potassium nitrate
Purity: 99.995% CAS No.: 7757-79-1 Molecular Formula: KNO3 Formula Weight: 101.11 Physical Form: Powder Density: 2.109 Appearance: White Odour: Odourless Melting Point: 334° Boiling Point: 400° dec. Merck Reference: 14,7648 UN No.: 1486 MDL No.: MFCD00011409
Formule: KNO₃
MW: 101.1 g/mol
Kookpunt: 400 °C (ontbinding)
Smeltpunt: 334
Dichtheid: 2.109
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011409
CAS nummer: 7757-79-1
EINECS: 231-818-8
UN: 1486
ADR: 5.1 III
Bereik: 01-2119488224-35
Merck Index: 14,7648