Lithium nitrate 99+%, extra pure
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
211320010 211322500 211320050
ACRO211320010EA 254 EUR
ACRO211320010 ACRO211322500 ACRO211320050
Lithium nitrate 99+%, extra pure
Lithium nitrate
Formule: LiNO₃
MW: 68.95 g/mol
Smeltpunt: 251 °C
Dichtheid: 2.38 g/cm³
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011094
CAS nummer: 7790-69-4
EINECS: 232-218-9
UN: 2722
ADR: 5.1 III
Merck Index: 13,05558
Order Now
Specification Test Results
Appearance White Adhering crystalline powder
Loss on drying ≤1 % (200°C)
Titration after Ion exchange ≥99.0 %