Cover glasses for fluorescence
Leverancier: MARIENFELD
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0100192 0100242 0102222 0102242 0103192 0103222 0103242 0100222 0100122 0102192 0102122 0102172 0102112 0102152 0100172
MARI0100192EA 85.3 EUR
MARI0100192 MARI0100242 MARI0102222 MARI0102242 MARI0103192 MARI0103222 MARI0103242 MARI0100222 MARI0100122 MARI0102192 MARI0102122 MARI0102172 MARI0102112 MARI0102152 MARI0100172
Cover glasses for fluorescence
Dekglaasjes
These cover glasses are made of borosilicate glass D 263™ M of the hydrolytic class 1 and are suitable for fluorescence microscopy.
  • Colourless and clear
  • Chemically resistant
  • Approved for in vitro diagnostic (IVD) applications according to IVD directive 98/79/EC
ISO 8255
Verpakking: In hinged lid plastic boxes of 100 or 200 pieces - Ten boxes in a carton.