Hose connectors, without nut
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
D568-14 D569-14 D568-25 D569-18 D568-32 D568-18
BOHLD568-14EA 6.3 EUR
BOHLD568-14 BOHLD569-14 BOHLD568-25 BOHLD569-18 BOHLD568-32 BOHLD568-18
Hose connectors, without nut
Fittingen en connectoren
GL 14 and GL 18 hose connectors made of PFA, GL 25 and GL 32 hose connectors made of PTFE with elastic sealing lip and FPM O-ring.
  • Available as straight or bent type
  • Universal chemical resistance
  • Maximum temperature up to 200 °C