Large microscopy slides
Leverancier: MARIENFELD
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1100420 1100220 1100020
MARI1100420EA 50.6 EUR
MARI1100420 MARI1100220 MARI1100020
Large microscopy slides
Object- en dekglaasjes Objectglaasjes
Soda lime glass microscopy slides precleaned. Also available with a silky frosted marking area of 20 mm on both sides.
Certificaten: IVD directive 98/79/EC.