GL tube fittings, ground joint, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
D570-32 D570-08 D570-16
BOHLD570-32EA 44.6 EUR
BOHLD570-32 BOHLD570-08 BOHLD570-16
GL tube fittings, ground joint, BOLA
Adapters Adapters met NS-verbindingen
PTFE, white. For connecting tubes or tubing to vessels with ground joint. For inserting and fixing probes, thermometers, dip tubes or cables.
  • Temperature resistance: −200 to +250 °C
  • Universal chemical resistance
  • Vacuum: suitable
Fitting made of PTFE, for transition from ground joints to GL threads. For connection of hard-walled tubing (e.g. PTFE, PFA, FEP) with BOLA Laboratory Screw Joints . Ground joint body with turned rings and knurled grip for opening. The product is only exposed to PTFE.