Socket GL tube fittings, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
D571-32 D571-16 D571-08
BOHLD571-32EA 60.6 EUR
BOHLD571-32 BOHLD571-16 BOHLD571-08
Socket GL tube fittings, BOLA
Adapters Adapters met NS-verbindingen
PTFE, white.
  • Temperature resistance: −200 to +250 °C
  • Universal chemical resistance
  • Vacuum: suitable
Fitting with socket for tubes with ground joint and GL thread for connection of hard-walled tubing (e.g. PTFE, PFA, FEP) with laboratory screw joints. The products is only exposed to PTFE.