Spherical ground joint GL tube fittings, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
D790-24 D790-48 D790-36
BOHLD790-24EA 31.8 EUR
BOHLD790-24 BOHLD790-48 BOHLD790-36
Spherical ground joint GL tube fittings, BOLA
Adapters Adapters met NS-verbindingen
PTFE, white. For connecting tubes or tubing to vessels with ground joint. For inserting and fixing probes, thermometers, dip tubes or cables.
  • Temperature resistance: −200 to +250 °C
  • Universal chemical resistance
  • Vacuum: suitable
Fitting for transition from spherical ground joints to GL threads. For connection of hard-walled tubing (e.g. PTFE, PFA, FEP) with laboratory screw joints. The product is only exposed to PTFE.