Multiple distributors for bottles, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
D612-08 D613-08
BOHLD612-08EA 103 EUR
BOHLD612-08 BOHLD613-08
Multiple distributors for bottles, BOLA
Sluitingen Flesdoppen
Green screw cap, made of PP, for GL 45 threaded bottles and distributor body made of PP.
  • Very good chemical resistance
  • Temperature resistant from 0 to +110 °C
  • Suitable for vacuum applications
For the removal or filling of aggressive or pure liquids. For the connection of hoses, tubes and probes into vessels. Hoses with a maximum external diameter of  8,5 mm can be inserted through the necks.