Staining jars acc. to Hellendahl
Leverancier: MARIENFELD
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
4200000 4200001
631-0698EA 247 EUR
631-0698 MARI4200001
Staining jars acc. to Hellendahl
Potjes Kleurbakjes
Jars, made of soda-lime glass, with lid.
These staining trough hold up to 8 slides (76×26 mm).
Jars made of heavy-walled soda lime glass with a relatively high coefficient of expansion, and it should not exposed to rapid temperature changes, to avoid cracks caused by tensions. Borosilicate glass jars are suitable for dish washers and microwave ovens.