Staining jars
Leverancier: MARIENFELD
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
6611000 4200004 6618000 4200005
MARI6611000EA 32 EUR
MARI6611000 MARI4200004 MARI6618000 MARI4200005
Staining jars
Potjes Kleurbakjes
Staining set consists of glass dish and staining jar made of soda lime glass, and staining tray made of either pressed glass or stainless steel.
These staining tray hold up to 10 slides (76×26 mm).
Jars made of heavy-walled soda lime glass with a relatively high coefficient of expansion, and it should not exposed to rapid temperature changes, to avoid cracks caused by tensions.