Staining jars according to Hausser/Gedigk
Leverancier: MARIENFELD
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
4200012 4200013 6618005
MARI4200012EA 45.9 EUR
MARI4200012 MARI4200013 631-9210
Staining jars according to Hausser/Gedigk
Potjes Kleurbakjes
Staining set consists of staining jar and glass dish made of soda lime glass (pressed glass), and staining tray made of stainless steel.
  • Cover is made of soda lime glass, with frosted rim of 10 mm width
Staining tray hold up to 60 slides (76×26 mm).
Order Now
Specificaties
Environmentally Preferable