GL Quick tube connectors, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
D625-20 D625-60 D625-40
BOHLD625-20EA 1150 EUR
BOHLD625-20 BOHLD625-60 BOHLD625-40
GL Quick tube connectors, BOLA
Fittingen en connectoren
Two-part quick connector is made of PFA, with two GL threads. When disconnected, the flow is interrupted by means of built-in non-return valves.
  • Suitable for pressure up to max. 6 bar
  • Universal chemical resistance
  • Maximum temperature up to 250 °C