Multiple distributors for bottles, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
D750-24 D750-16
BOHLD750-24EA 125 EUR
BOHLD750-24 BOHLD750-16
Multiple distributors for bottles, BOLA
Sluitingen Flesdoppen
Screw cap and distributor body both made of PP.
  • Very good chemical resistance
  • Temperature resistant from 0 to +110 °C
  • Suitable for vacuum applications
For the removal or filling of aggressive or pure liquids. For the connection of hoses, tubes and probes into vessels. Hoses with different external diameters can be inserted through the necks.