Distributors for reaction vessels (R), BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
D744-24 D744-16
BOHLD744-24EA 328 EUR
BOHLD744-24 BOHLD744-16
Distributors for reaction vessels (R), BOLA
Adapters Adapters met NS-verbindingen
Blue screw cap made of PP with GLS 80 thread, body made of PTFE.
  • Very good chemical resistance
  • For working temperatures up to max. +200 °C (PP screw cap max. +110 °C).
Center neck with GL 25 thread for connecting tubing or tube up to a max. O.D. of 15 mm, four lateral necks with GL 18 thread. Exchangeable shaft guide (PTFE) with screw cap (PPS) for inserting and fixing a stirrer shaft in the center neck included.