Distributors for reaction vessels (S), BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
D748-16 D748-60 D748-40
BOHLD748-16EA 332 EUR
BOHLD748-16 BOHLD748-60 BOHLD748-40
Distributors for reaction vessels (S), BOLA
Sluitingen Flesdoppen
PP screw cap, blue and PTFE body.
  • Very good chemical reistance
  • For working temperatures up to max. +250 °C (pp screw cap max. +140 °C)
  • FDC conform
Screw cap with GLS 80® thread, body with center ground joint and lateral GL-threaded or ground joint necks. Applications: Drawing or inserting aggressive or pure liquids. Inserting tubing, tubes and probes into vessels. Use of a stirrer bearing in center neck for centrical position of a stirrer shaft.