Celtellers met drie rasters, McMaster
Leverancier: MARIENFELD
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0611141 0335001
631-0695EA 391 EUR
631-0695 MARI0335001
Celtellers met drie rasters, McMaster
Object- en dekglaasjes Telkamers
This cell counter is suitable for liquid media.
  • Three counting grids and spare cover glass
  • Pair of clamps to fix the cover glass
  • Ground edges and rounded corners