Flexible distributors for bottles, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
D800-48 D800-24 D800-36
201-1087EA 74.7 EUR
201-1087 201-1085 201-1086
Flexible distributors for bottles, BOLA
Sluitingen Flesdoppen
PP screw cap, green, distributor body and tubing connectors made from PP
  • Very good chemical resistance
  • Temperature resistance from 0 to +110 °C
  • Suitable for vacuum
Drawing or inserting liquids. For elastic tubing (e.g. Viton®, Tygon®, silicone). Bent hose connectors on upper side, straight hose connectors on lower side.