Gaskets for screw caps, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
H973-18 H973-41
BOHLH973-18EA 1.8 EUR
BOHLH973-18 BOHLH973-41
Gaskets for screw caps, BOLA
Afdichtingen Pakkingen
Upper side of the screw caps is made of PTFE and lower side is made of silicone-elastomer for balancing unevennesses on sealing surfaces.
  • Universal chemical resistance
  • Temperature resistance: from −60 to +230 °C
After assembly, the product is only exposed to PTFE.